Anti-CD244 (2B4) Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: C1.7]

Supplier: Biolegend
329511 329512
CA329511-BLEA 237.77 CAD
CA329511-BL CA329512-BL
Anti-CD244 (2B4) Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: C1.7]
Antibodies
Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. Test size products are transitioning from 20 μl to 5 μl per test. Please check your vial or your CoA to find the suggested use of this reagent per million cells in 100 μl staining volume or per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD244 (2B4)
Clonality: Monoclonal
Clone: C1.7
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR