Anti-CD209 (DC-SIGN) Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 9E9A8]

Supplier: Biolegend
330107 330108
CA330107-BLEA 237.77 CAD
CA330107-BL CA330108-BL
Anti-CD209 (DC-SIGN) Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 9E9A8]
Antibodies
APC anti-human CD209 (DC-SIGN) [9E9A8]; Isotype: Mouse IgG2a, κ; Reactivity: Human; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. Test size products are transitioning from 20 μl to 5 μl per test. Please check your vial or your CoA to find the suggested use of this reagent per million cells in 100 μl staining volume or per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application. Read more at www.biolegend.com/testsize regarding the test size change.

Type: Primary
Antigen: CD209 (DC-SIGN)
Clonality: Monoclonal
Clone: 9E9A8
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: Mouse IgG2a, κ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR