Anti-CD137 (4-1BB) Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 4B4-1]
Supplier: Biolegend
309809 309810
CA309809-BLEA 229.8 CAD
CA309809-BL CA309810-BL
Anti-CD137 (4-1BB) Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 4B4-1]
Antibodies
APC anti-human CD137 (4-1BB) [4B4-1]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Chimpanzee, Baboon, Cynomolgus, Rhesus; Apps: FC; Size: 100 tests
Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. Test size products are transitioning from 20 μl to 5 μl per test. Please check your vial or your CoA to find the suggested use of this reagent per million cells in 100 μl staining volume or per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application. ReadMore Product Information