Anti-CD4 Mouse Monoclonal Antibody (Alexa Fluor® 700) [clone: OKT4]

Supplier: Biolegend
317425 317426
CA317425-BLEA 197.88 CAD
CA317425-BL CA317426-BL
Anti-CD4 Mouse Monoclonal Antibody (Alexa Fluor® 700) [clone: OKT4]
Antibodies
Alexa Fluor® 700 anti-human CD4 [OKT4]; Isotype: Mouse IgG2b, κ; Reactivity: Human, Cross-Reactivity: Chimpanzee, Cynomolgus, Rhesus; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. The suggested use of this reagent is 1.0 μg per million cells in 100 μl volume. It is highly recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD4
Clonality: Monoclonal
Clone: OKT4
Conjugation: Alexa Fluor® 700
Epitope:
Host: Mouse
Isotype: Mouse IgG2b, κ
Reactivity: Human;Cross-Reactivity: Chimpanzee;Cynomolgus;Rhesus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR