Anti-Notch 3 Armenian Hamster Monoclonal Antibody (Alexa Fluor® 647) [clone: HMN3-133]

Supplier: Biolegend
130511 130512
CA130511-BLEA 234.58 CAD
CA130511-BL CA130512-BL
Anti-Notch 3 Armenian Hamster Monoclonal Antibody (Alexa Fluor® 647) [clone: HMN3-133]
Antibodies
Alexa Fluor® 647 anti-mouse Notch 3 [HMN3-133]; Isotype: Armenian Hamster IgG; Reactivity: Mouse; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 1.0 μg per 106 cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

* Alexa Fluor® 647 has a maximum emission of 668 nm when it is excited at 633nm / 635nm.

Type: Primary
Antigen: Notch 3
Clonality: Monoclonal
Clone: HMN3-133
Conjugation: Alexa Fluor® 647
Epitope:
Host: Armenian Hamster
Isotype: Armenian Hamster IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR