Anti-Ly-6G/Ly-6C (Gr-1) Rat Monoclonal Antibody (Alexa Fluor® 647) [clone: RB6-8C5]

Supplier: Biolegend
108418 108420
CA108418-BLEA 365.44 CAD
CA108418-BL CA108420-BL
Anti-Ly-6G/Ly-6C (Gr-1) Rat Monoclonal Antibody (Alexa Fluor® 647) [clone: RB6-8C5]
Antibodies
Alexa Fluor® 647 anti-mouse Ly-6G/Ly-6C (Gr-1) [RB6-8C5]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC; Size: 25 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per 106 cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: Ly-6G/Ly-6C (Gr-1)
Clonality: Monoclonal
Clone: RB6-8C5
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rat
Isotype: Rat IgG2b, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR