Anti-I-Ad Mouse Monoclonal Antibody (Alexa Fluor® 647) [clone: 39-10-8]

Supplier: Biolegend
115009 115010
CA115009-BLEA 161.18 CAD
CA115009-BL CA115010-BL
Anti-I-Ad Mouse Monoclonal Antibody (Alexa Fluor® 647) [clone: 39-10-8]
Antibodies
Alexa Fluor® 647 anti-mouse I-Ad [39-10-8]; Isotype: Mouse (C3H.SW) IgG3, κ; Reactivity: Mouse d haplotype, Does not cross-react with other haplotypes (e.g., a, b, k, p, q, s); Apps: FC, IHC, IF; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: I-Ad
Clonality: Monoclonal
Clone: 39-10-8
More Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR