Anti-CD49e Rat Monoclonal Antibody (Alexa Fluor® 647) [clone: 5H10-27(MFR5)]

Supplier: Biolegend
103811 103812
CA103811-BLEA 161.18 CAD
CA103811-BL CA103812-BL
Anti-CD49e Rat Monoclonal Antibody (Alexa Fluor® 647) [clone: 5H10-27(MFR5)]
Antibodies
Alexa Fluor® 647 anti-mouse CD49e [5H10-27(MFR5)]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC, IF, IHC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 1.0 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD49e
Clonality: Monoclonal
Clone: 5H10-27(MFR5)
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rat
Isotype: Rat IgG2a, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR