Anti-CD4 Rat Monoclonal Antibody (Alexa Fluor® 647) [clone: GK1.5]

Supplier: Biolegend
100424 100426
CA100424-BLEA 365.44 CAD
CA100424-BL CA100426-BL
Anti-CD4 Rat Monoclonal Antibody (Alexa Fluor® 647) [clone: GK1.5]
Antibodies
Alexa Fluor® 647 anti-mouse CD4 [GK1.5]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC, IHC, IF; Size: 25 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per million cells in 100 μl volume. For immunohistochemisty, a concentration range of 2.5-5 μg/ml is suggested. For immunofluorescence microscopy, a concentration range of 1.25-10 μg/ml is recommended. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD4
Clonality: Monoclonal
Clone: GK1.5
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rat
Isotype: Rat IgG2b, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR