Anti-CD3 epsilon Armenian Hamster Monoclonal Antibody (Alexa Fluor® 647) [clone: 145-2C11]

Supplier: Biolegend
100322 100324
CA100322-BLEA 365.44 CAD
CA100322-BL CA100324-BL
Anti-CD3 epsilon Armenian Hamster Monoclonal Antibody (Alexa Fluor® 647) [clone: 145-2C11]
Antibodies
Alexa Fluor® 647 anti-mouse CD3ε [145-2C11]; Isotype: Armenian Hamster IgG; Reactivity: Mouse; Apps: FC, IHC-F, IF; Size: 25 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per million cells in 100 μl volume. For immunohistochemistry, a concentration range of 5.0 - 10.0 μg/ml is suggested. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD3epsilon
Clonality: Monoclonal
Clone: 1More Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR