Anti-SSEA-4 Mouse Monoclonal Antibody (Alexa Fluor® 647) [clone: MC-813-70]

Supplier: Biolegend
330407 330408
CA330407-BLEA 263.31 CAD
CA330407-BL CA330408-BL
Anti-SSEA-4 Mouse Monoclonal Antibody (Alexa Fluor® 647) [clone: MC-813-70]
Antibodies
Alexa Fluor® 647 anti-human SSEA-4 [MC-813-70]; Isotype: Mouse IgG3, κ; Reactivity: Human, Baboon, Cynomolgus, Mouse, Rhesus; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood.  It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: SSEA-4
Clonality: Monoclonal
Clone: MC-813-70
Conjugation: Alexa Fluor® 647
Epitope:
Host: Mouse
Isotype: Mouse IgG3, κ
Reactivity: Human;Baboon;Cynomolgus;Mouse;Rhesus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR