Anti-CD61 Mouse Monoclonal Antibody (Alexa Fluor® 647) [clone: VI-PL2]

Supplier: Biolegend
336407 336408
CA336407-BLEA 225.01 CAD
CA336407-BL CA336408-BL
Anti-CD61 Mouse Monoclonal Antibody (Alexa Fluor® 647) [clone: VI-PL2]
Antibodies
Alexa Fluor® 647 anti-human CD61 [VI-PL2]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cynomolgus, Rhesus; Apps: FC, IF; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells in 100 μl volume or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD61
Clonality: Monoclonal
Clone: VI-PL2
Conjugation: Alexa Fluor®More Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR