Anti-CD209 (DC-SIGN) Mouse Monoclonal Antibody (Alexa Fluor® 647) [clone: 9E9A8]

Supplier: Biolegend
330111 330112
CA330111-BLEA 197.88 CAD
CA330111-BL CA330112-BL
Anti-CD209 (DC-SIGN) Mouse Monoclonal Antibody (Alexa Fluor® 647) [clone: 9E9A8]
Antibodies
Alexa Fluor® 647 anti-human CD209 (DC-SIGN) [9E9A8]; Isotype: Mouse IgG2a, κ; Reactivity: Human; Apps: FC, IF; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD209 (DC-SIGN)
Clonality: Monoclonal
Clone: 9E9A8
Conjugation: Alexa Fluor® 647
Epitope:
Host: Mouse
Isotype: Mouse IgG2a, κ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR