Anti-CD14 Mouse Monoclonal Antibody (Alexa Fluor® 647) [clone: M5E2]

Supplier: Biolegend
301812 301818
CA301812-BLEA 422.89 CAD
CA301812-BL CA301818-BL
Anti-CD14 Mouse Monoclonal Antibody (Alexa Fluor® 647) [clone: M5E2]
Antibodies
Alexa Fluor® 647 anti-human CD14 [M5E2]; Isotype: Mouse IgG2a, κ; Reactivity: Human, African Green, Capuchin Monkey, Cattle (Bovine, Cow), Chimpanzee, Common Marmoset, Cotton-topped Tamarin, Cynomolgus, Dog (Canine), Rhesus, Pigtailed Macaque, Squirrel Monkey; Apps: FC; Size: 25 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD14
Clonality: Monoclonal
Clone: M5E2
Conjugation: Alexa Fluor® 647
Epitope:
Host: Mouse
Isotype: Mouse IgG2a, κ
Reactivity: Human;African Green;Capuchin Monkey;Cattle (Bovine;Cow);Chimpanzee;Common Marmoset;Cotton-topped Tamarin;Cynomolgus;Dog (Canine);Rhesus;Pigtailed Macaque;Squirrel Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR