Anti-Panendothelial Cell Antigen Rat Monoclonal Antibody (Alexa Fluor® 488) [clone: MECA-32]

Supplier: Biolegend
120505 120506
CA120505-BLEA 161.18 CAD
CA120505-BL CA120506-BL
Anti-Panendothelial Cell Antigen Rat Monoclonal Antibody (Alexa Fluor® 488) [clone: MECA-32]
Antibodies
Alexa Fluor® 488 anti-mouse Panendothelial Cell Antigen [MECA-32]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: Panendothelial Cell Antigen
Clonality: Monoclonal
Clone: MECA-32
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rat
Isotype: Rat IgG2a, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR