Anti-CD80 Armenian Hamster Monoclonal Antibody (Alexa Fluor® 488) [clone: 16-10A1]

Supplier: Biolegend
104715 104716
CA104715-BLEA 161.18 CAD
CA104715-BL CA104716-BL
Anti-CD80 Armenian Hamster Monoclonal Antibody (Alexa Fluor® 488) [clone: 16-10A1]
Antibodies
Alexa Fluor® 488 anti-mouse CD80 [16-10A1]; Isotype: Armenian Hamster IgG; Reactivity: Mouse, Cross-Reactivity: Dog (Canine); Apps: FC, IHC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 2.0 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD80
Clonality: Monoclonal
Clone: 16-10A1
Conjugation: Alexa Fluor® 488
Epitope:
Host: Armenian Hamster
Isotype: Armenian Hamster IgG
Reactivity: Mouse;Cross-Reactivity: Dog (Canine)
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR