Anti-CD45.1 Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: A20]

Supplier: Biolegend
110717 110718
CA110717-BLEA 161.18 CAD
CA110717-BL CA110718-BL
Anti-CD45.1 Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: A20]
Antibodies
Alexa Fluor® 488 anti-mouse CD45.1 [A20]; Isotype: Mouse (A.SW) IgG2a, κ; Reactivity: Mouse; Apps: FC, IHC, IF; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per 106 cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD45.1
Clonality: Monoclonal
Clone: A20
Conjugation: Alexa Fluor® 488
Epitope:
Host: Mouse
Isotype: Mouse (A.SW) IgG2a, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR