Anti-CD4 Rat Monoclonal Antibody (Alexa Fluor® 488) [clone: GK1.5]
Supplier: Biolegend
100423 100425
CA100423-BLEA 365.44 CAD
CA100423-BL CA100425-BL
Anti-CD4 Rat Monoclonal Antibody (Alexa Fluor® 488) [clone: GK1.5]
Antibodies
Alexa Fluor® 488 anti-mouse CD4 [GK1.5]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC, IHC, IF; Size: 25 μg
Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.06 μg per million cells in 100 μl volume. For immunohistochemistry, a concentration range of 2.5-5 μg/ml is suggested. For immunofluorescence microscopy, a concentration range of 1.25-10 μg/ml is recommended. It is recommended that the reagent be titrated for optimal performance for eachMore Product Information