Anti-CD11c Armenian Hamster Monoclonal Antibody (Alexa Fluor® 488) [clone: N418]

Supplier: Biolegend
117311 117313
CA117311-BLEA 365.44 CAD
CA117311-BL CA117313-BL
Anti-CD11c Armenian Hamster Monoclonal Antibody (Alexa Fluor® 488) [clone: N418]
Antibodies
Alexa Fluor® 488 anti-mouse CD11c [N418]; Isotype: Armenian Hamster IgG; Reactivity: Mouse; Apps: FC, IHC, IF; Size: 25 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per 106 cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for other applications.

Type: Primary
Antigen: CD11c
Clonality: Monoclonal
Clone: N418
Conjugation: Alexa Fluor® 488
Epitope:
Host: Armenian Hamster
Isotype: Armenian Hamster IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR