Anti-SSEA-3 Rat Monoclonal Antibody (Alexa Fluor® 488) [clone: MC-631]

Supplier: Biolegend
330305 330306
CA330305-BLEA 263.31 CAD
CA330305-BL CA330306-BL
Anti-SSEA-3 Rat Monoclonal Antibody (Alexa Fluor® 488) [clone: MC-631]
Antibodies
Alexa Fluor® 488 anti-human/mouse SSEA-3 [MC-631]; Isotype: Rat IgM, κ; Reactivity: Human, African Green, Mouse; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: SSEA-3
Clonality: Monoclonal
Clone: MC-631
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rat
Isotype: Rat IgM, κ
Reactivity: Human;African Green;Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR