Anti-CD45 Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: HI30]

Supplier: Biolegend
304017 304019
CA304017-BLEA 386.18 CAD
CA304017-BL CA304019-BL
Anti-CD45 Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: HI30]
Antibodies
Alexa Fluor® 488 anti-human CD45 [HI30]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Chimpanzee; Apps: FC; Size: 25 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD45
Clonality: Monoclonal
Clone: HI30
Conjugation: Alexa Fluor® 488
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;Cross-Reactivity: Chimpanzee
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR