Anti-TAF1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
600-401-995
CA600-401-995EA 834 CAD
CA600-401-995
Anti-TAF1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: TAF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR