Mouse Recombinant EXODUS-2/CCL21 (from E. coli)
Supplier: FUJIFILM IRVINE SCIENTIFIC, INC
200-35-1MG 200-35-20UG 200-35-5UG 200-35-100UG 200-35-500UG
10788-084EA 6375.71 CAD
10788-084 10788-086 10788-088 10789-158 10821-938
Mouse Recombinant EXODUS-2/CCL21 (from E. coli)
Proteins and Peptides
Interferon gamma-induced protein 10 (IP-10), or CXCL10, is a chemokine secreted by monocytes, endothelial cells, and fibroblasts in response to interferon gamma (IFN-γ). IP-10 functions as a chemoattractant for activated T cells, monocytes, dendritic, and natural killer (NK) cells that express the G protein-coupled receptor CXCR3. IP-10 is an important factor in autoimmune diseases such as Hashimoto's thyroiditis, Graves' disease, and Type 1 diabetes mellitus.
  • High quality
  • Low endotoxin
  • Affordable
Shenandoah Biotechnology is a manufacturer of research grade recombinant proteins typically used as cytokines, growth factors and chemokines in cell based assays and cell culture. Each production lot is tested for QA and the expected values for each product can be viewed under the product specifications tab.
Caution: For research use only.