Mouse Recombinant LIF (from E. coli)
Supplier: FUJIFILM IRVINE SCIENTIFIC, INC
200-33-100UG 200-33-1MG 200-33-20UG 200-33-5UG 200-33-500UG
10788-066EA 1142.17 CAD
10788-066 10788-068 10788-070 10788-072 10822-242
Mouse Recombinant LIF (from E. coli)
Proteins and Peptides
Leukemia inhibitory factor (LIF) is a member of the interleukin 6 (IL-6) family that is made by a variety of adult and embryonic tissues. LIF signals through the glycoprotein 130 (gp130)/LIF receptor (LIFR) heterodimer to activate STAT3 and MAPK signaling. LIF functions during hematopoietic differentiation, neuronal cell differentiation, kidney development, and inflammatory processes. Mouse LIF promotes mouse embryonic stem (ES) cell self-renewal and pluripotency in long-term cell culture systems, similar to the functional activity of FGF-basic in human ES cell culture systems.
  • High Quality
  • Low Endotoxin
  • Affordable
A COA is supplied with each product shipped detailing purity, formulation, biological activity, endotoxin levels, reconstitution instructions and recommended storage conditions.  This product is for research use only and is not for use in humans.  
Caution: For research use only.