Mouse Recombinant TPO (from E. coli)
Supplier: FUJIFILM IRVINE SCIENTIFIC, INC
200-32-100UG 200-32-10UG 200-32-1MG 200-32-500UG
10788-050EA 1145.07 CAD
10788-050 10788-052 10788-054 10788-056 10822-408
Mouse Recombinant TPO (from E. coli)
Proteins and Peptides
Thrombopoietin (TPO) is a growth factor that is produced by liver and kidney tissues. TPO binds the TPO receptor (CD110) to promote megakaryocyte maturation, differentiation, and the production of platelets.
  • High quality
  • Low endotoxin
  • Affordable
Shenandoah Biotechnology is a manufacturer of research grade recombinant proteins typically used as cytokines, growth factors and chemokines in cell based assays and cell culture. Each production lot is tested for QA and the expected values for each product can be viewed under the product specifications tab.
Caution: For research use only.