Mouse Recombinant TNF-A (from E. coli)
Supplier: FUJIFILM IRVINE SCIENTIFIC, INC
200-31-1MG 200-31-20UG 200-31-5UG 200-31-100UG 200-31-500UG
10788-044EA 5854.52 CAD
10788-044 10788-046 10788-048 10788-786 10822-400
Mouse Recombinant TNF-A (from E. coli)
Proteins and Peptides
Tumor necrosis factor alpha (TNF-α) is an inflammatory cytokine secreted by macrophages, monocytes, neutrophils, T cells, and NK-cells following stimulation by bacterial lipopolysaccharide (LPS). TNF-α signal activation occurs through two receptors, TNFR1 and TNFR2. TNFR1 is expressed on most cell types, unlike TNFR2, which is expressed mainly on immune cells. TNF-α functions to stimulate phagocytosis in macrophages, chemoattract neutrophils, increase insulin resistance, and induce fever.
  • High quality
  • Low endotoxin
  • Affordable
Caution: For research use only.