Dosing Pumps, Dosi-Pump, Bürkle
Supplier: BURKLE INC
5607-3030 5607-4100 5607-5250 5607-1004 5607-2030
10790-746EA 93.71 CAD
10790-746 10790-748 10790-750 75785-120 75785-122
Dosing Pumps, Dosi-Pump, Bürkle
Pumps Drum/Transfer Pumps
Dosi pump are hand controlled pump used for the controlled product extraction from canisters, bottles, barrels and carboys
  • Medium contacting parts made of PP, PE and stainless steel
  • Suitable for products of different viscosity
Dosi pump is inserted into container closures with suitable holes. Immersion value represents the value of Immersion depth.