Disposable Samplers, ZoneDispo, Burkle
Supplier: Bürkle
5393-4431 5393-4420 5393-4430 5393-4421
10769-584PK 923.95 CAD
10821-283 10821-243 10821-245 10769-584
Disposable Samplers, ZoneDispo, Burkle
Samplers Disposable Samplers
ZoneDispo disposable all-layer sampler is used in sampling to collect a representative all-layer sample from three different layers.
  • HDPE
  • Clean room manufactured
  • Optionally sterilised by gamma rays
Suitable for free-flowing powders and small-grain granulates. The collected sample can be secured and transported in the closed sampler.