LaboPlast® and SteriPlast® Bio Samplling Spatulas, Bürkle
Supplier: BURKLE INC
5379-1009 5379-0009
10769-680PK 72.77 CAD
10769-680 10769-684
LaboPlast® and SteriPlast® Bio Samplling Spatulas, Bürkle
Spatulas
These disposable spatulas are manufactured from bio-plastic 'Green PE' which is entirely made of renewable resources.
  • 'Green PE' is 100% recyclable, made without fossil raw materials
  • Long, ergonomic and solid handle
  • Pointed blade can be easily pierced into paper and plastic bags
  • Available non sterile (LaboPlast Bio) or gamma radiation sterilised (SteriPlast Bio)
  • Produced in a cleanroom environment
They are ideal for the sampling of powder, granules and pastes. Insertion depth of this bio sampling spatulas is 150 mm.