LaboPlast® and SteriPlast® Sampling Spatulas, Bürkle
Supplier: BURKLE INC
5378-1009 5378-0009 5378-1010 5378-0010
10769-678PK 61.79 CAD
10769-678 10769-682 10769-916 10769-918
LaboPlast® and SteriPlast® Sampling Spatulas, Bürkle
Spatulas
These disposable spatulas are ideal for the sampling of powder, granules and pastes.
  • Long, ergonomic and solid handle
  • Pointed blade can be easily pierced into paper and plastic bags
  • Available non sterile (LaboPlast) or gamma radiation sterilised (SteriPlast)
  • Production, assembling and packaging acc. to ISO cleanroom class 7
  • According to FDA guidelines
As an option, the spatulas are available with cover in order to secure a contamination-free sampling process. Insertion depth of LaboPlast and SteriPlast spatula is 150 mm.