VWR® Round Carboy with Spigot, Narrow Mouth
Supplier: VWR International
10755-088 10755-090 10755-116 10755-118 10755-128 10755-130 75795-676 75795-682 75795-698
10755-088CS 1775.66 CAD
10755-088 10755-090 10755-116 10755-118 10755-128 10755-130 75795-676 75795-682 75795-698
VWR® Round Carboy with Spigot, Narrow Mouth
Bottles Carboys
Use for storing and dispensing solutions and media. Ideal for sterile water storage.
  • Non sterile
  • Polypropylene, LDPE, and HDPE formats available
  • Excellent chemical resistance
  • Clear graduation marks
  • Narrow-mouth
  • Feature an integrated shoulder handle
Carboys are well suited for use in temperatures ranging from -100 to 100 °C (-148 to 212 °F). Carboys are leak-proof on standard test conditions. Spigots are tied to the handle, and require assembly by hand for usage. No tools are necessary for assembly.
Polypropylene carboys can be autoclaved beforeMore Product Information