VWR® Heavy-Duty Round Carboy, Narrow Mouth
Supplier: VWR International
10755-092 10755-104
10755-092CS 1787.75 CAD
10755-092 10755-104
VWR® Heavy-Duty Round Carboy, Narrow Mouth
Bottles Carboys
Use for storing and dispensing solutions and media. Ideal for sterile water storage.
  • Heavy duty construction
  • Non sterile
  • Polypropylene formats available
  • Excellent chemical resistance
  • Clear graduation marks
  • Narrow-mouth
  • Feature an integrated shoulder handle
Carboys are well suited for use in temperatures ranging from –100 to 100 °C (–148 to 212 °F). Carboys are leak-proof under standard test conditions.
Polypropylene carboys can be autoclaved before refilling to stop recurring bacterial growth.
Certifications: Resin meets US FDA 21 CFR, USP Class VI.