VWR® Round Carboy, Wide Mouth
Supplier: VWR International
10755-100 10755-102 10755-122 75795-690
10755-100CS 900.54 CAD
10755-100 10755-102 10755-122 75795-690
VWR® Round Carboy, Wide Mouth
Bottles Carboys
Use for storing and dispensing solutions and media. Ideal for sterile water storage.
  • Non sterile
  • Polypropylene and LDPE formats available
  • Excellent chemical resistance
  • Clear graduation marks
  • Wide-mouth
  • Feature an integrated shoulder handle
Carboys are well suited for use in temperatures ranging from -100 to 100 °C (-148 to 212 °F). Carboys are leak-proof under standard test conditions.
Polypropylene carboys can be autoclaved before refilling to stop recurring bacterial growth. LDPE carboys are not autoclavable.
Certifications: Resin meets US FDAMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable