VWR® Rectangular Carboy with Spigot, Wide Mouth
Supplier: VWR International
683242 683250 683243 683260 583211
10755-094CS 1607.43 CAD
10755-094 10755-096 10755-106 10755-108 10766-762
VWR® Rectangular Carboy with Spigot, Wide Mouth
Bottles Carboys
Use for storing and dispensing solutions and media. Ideal for sterile water storage.
  • Non sterile
  • Polypropylene and HDPE formats available
  • Clear graduation marks
  • Excellent chemical resistance
  • Wide mouth
  • Heavy duty stainless steel handle for easy carrying and pouring
Carboys are well suited for use in temperatures ranging from –100 to 100 °C (–148 to 212 °F). Carboys are leak proof under standard test conditions. Spigots are tied to the handle, and require assembly by hand for usage. No tools are necessary for assembly.
The high-density polyethylene, disposable carboys are ideal for storage and transfer of sterile fluids and pharmaceutical/biotech reagents. Carboys offer excellent chemical resistance and are well suited for use from –100 to 100 °C (–148 to 212 °F). Graduated to identify the volume despensed.
Polypropylene carboys can be autoclaved before refilling to stop recurring bacterial growth. HDPE carboys are not autoclavable.
Certifications: Resin meets US FDA 21 CFR, USP Class VI.
Ordering information: Spigot included. Always disassemble spigot and cap from carboy before autoclaving.