LaboPlast® Scoop, Bürkle
Supplier: BURKLE INC
5378-0001 5378-0002 5378-0003 5378-0004 5378-0005 5378-0006 5378-0007 5378-0008 5378-0013 5378-0016 5378-0014 5378-0015 5378-0019 5378-5013 5378-5017 5378-0020
10769-670PK 63.45 CAD
10769-670 10769-672 10769-674 10769-676 10769-712 10769-714 10769-716 10770-180 10810-737 10810-759 10810-855 10810-857 75848-652 76501-358 76501-368 76501-394
LaboPlast® Scoop, Bürkle
Scoops
These scoops are ideal for the sampling of powder, granules and pastes.
  • Individually packed
  • Production, assembling and packaging acc. to ISO cleanroom class 7
They are also available with a transparent lid to close it immediately after the sampling procedure to secure a clean and contamination-free sample. Due to the special handle construction and the slightly angled edge the scoop stands horizontal and can be laid down without the material trickling out.
Certifications: Complies with EU food requirements and FDA regulations.