Unichillers, Huber
Supplier: Huber
3015.0062.01 3015.0064.01 3026.0113.01 3026.0114.01 3016.0025.01 3016.0028.01 3015.0072.01 3015.0073.01 3026.0107.01 3016.0131.01 3016.0033.01
10220-792EA 113332.04 CAD
10220-792 10220-794 10220-796 10220-798 10221-230 10221-232 10221-234 10221-236 10221-238 10221-240 10221-242 10221-244 10221-246 10221-248 10221-252 10221-254 10221-256 10221-258 10221-260 10221-262
Unichillers, Huber
Circulating Thermostats Closed Circulator Systems Cooling Circulators
Huber Unichillers offer better efficiency, stable pressure and delivery performance with a constant cooling water temperature compared to traditional tap water cooling
Unichillers have the following advantages and functions: environmentally-friendly, economic cooling solution, high efficiency, precise temperature control, powerful circulation pump, robust stainless construction, reliable continuously operation with warning functions, easy to fill and empty, illuminated filling level indicator, LED temperature display, easy operation with only three keys.
Chillers also reduce waterMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable