Multi-Use Test Tubes, Globe Scientific
Supplier: Globe Scientific
113010 118011 117011
10014-226PK 287.32 CAD
10014-226 10014-236 10014-250
Multi-Use Test Tubes, Globe Scientific
Tubes Test Tubes
Globe Scientific offers an extensive line of multi-use test tubes.
  • Polystyrene tubes
  • Withstand 1500 rpm during centrifugation
Tubes are molded from premium grade polystyrene for clarity. Tubes are designed to fit most laboratory applications.