Protégé, Nitrile Exam Gloves, Supermax Healthcare Canada
Supplier: Supermax Healthcare Canada
Aurelia
93995
CA10500-540CS 276.11 CAD
CA10500-540
Protégé, Nitrile Exam Gloves, Supermax Healthcare Canada
Gloves
Enhanced formulation rivals latex comfort and ergonomics.
  • Textured fingertips for precision gripping
  • Great resistance
  • Hypoallergenic
  • Each batch of gloves is air-tested to detect pinholes
  • Protect hands from biohazards and a broad range of chemicals
Certifications: ISO 9001: 2000 certified by SIRIM (Malaysia)
ISO 9001: 2000 and ISO 13485: 2003 certified by SGS (UK)
Ordering information: Packaged 100 gloves per dispenser box.