Mouse Recombinant Resistin (from E. coli)
Supplier: FUJIFILM IRVINE SCIENTIFIC, INC
Resistin, also known as FIZZ3, is a peptide hormone belonging to a class of cysteine-rich secreted proteins termed the resistin-like molecules (RELM) family. Mouse resistin, produced by adipocytes, is involved in insulin resistance and modulates glucose homeostasis and adipogenesis.
- High Quality
- Low Endotoxin
- Affordable
A COA is supplied with each product shipped detailing purity, formulation, biological activity, endotoxin levels, reconstitution instructions and recommended storage conditions. This product is for research use only and is not for use in humans.
Caution: For research use only.