False Bottom Tubes and Caps, Globe Scientific
Supplier: Globe Scientific
5527 5527B 5528B
10014-260CS 307.76 CAD
10014-260 10014-262 10014-268
False Bottom Tubes and Caps, Globe Scientific
Tubes Test Tubes
False bottom tubes are designed for use as a secondary tube for barcoding on automated analyzers.
Tubes may be used with a variety of popular analyzers, including Hitachi®, Olympus®, Corning®, Ortho®, Kodak®, Beckman®, IA®, Tosoh®, Vitros®, and more.