Water, WFI (water for injection) quality, sterile VAI®
Supplier: VELTEK ASSOCIATES INC
VAI-WFI-SP-11Z
75795-392CS
1134.44
CAD
75795-392
Water, WFI (water for injection) quality, sterile VAI®
Water
VAI WFI Quality water is a high quality water produced from validated WFI water system used for disinfectant preparation and dilution not for parenteral administration.
- Filtered at 0.2 micron
- Tested for assay, sterility and endotoxin levels
- Manufactured in a GMP, Class 100 (ISO 5, Grade A) area
- Gamma irradiated and double bagged packaged
Delivered with lot specific Certificate of Analysis and Sterility Report.
Formula:
H₂O Boiling Pt: 100 °C (1013 hPa) Melting Pt: 0 °C Density: 1 g/cm³ (4 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011332 CAS Number: 7732-18-5 EINECS: 231-791-2 Merck Index: 13,10098 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...