Diffraction Grating
Supplier: RAINBOW SYMPHONY INC SE
01604/15
470191-290KT 26.4 CAD
470191-290
Diffraction Grating
Educational Materials Physics Educational Materials Optics Learning Activities
Slides measure 2 x 2", with 500 lines per mm.