OptiTDS™, Reproductive Tissue Dissociation, CHI Scientific
Supplier: CHI Scientific
4-20411 4-20412 4-20413 4-20414 4-20415 4-20416 4-20417 4-20418 4-20419 4-20420 4-20421 4-20422 4-20423 4-20424 4-20425 4-20426 4-20427 4-20428 4-20429
10783-256EA 538.12 CAD
10783-256 10783-258 10783-260 10783-262 10783-264 10783-266 10783-268 10783-270 10783-272 10783-274 10783-276 10783-278 10783-280 10783-282 10783-284 10783-286 10783-288 10783-290 10783-292
OptiTDS™, Reproductive Tissue Dissociation, CHI Scientific
Protein Extraction Protein Extraction Kits
OptiTDS™ is a proprietary Tissue Dissociation system which enzymatically digests extracellular connective fibers without harming the cells themselves
Stability/Storage: The reagents contained in this system are stable at room temperatures for normal shipping periods, but the kit should be used immediately or the package should be stored at -20oC upon arrival. The reagents contained in this kit are stable and effective for up to 6 months at -20oC.
Each OptiTDS™ system contains concentrated tissue dissociation enzymes and reagents which are added to an included buffer system to produce a working solution that is capable of treating 3-5 g of tissue. Each tissue dissociation system is customized to be tissue-specific and cell-specific to obtain the best results for each type of primary cell.