OptiTDS™, Neural Tissue Dissociation System, CHI Scientific
Supplier: CHI Scientific
4-20351 4-20352 4-20353 4-20354 4-20355 4-20356 4-20357 4-20358 4-20359 4-20360 4-20361 4-20362
10783-202EA 601.46 CAD
10783-202 10783-228 10783-230 10783-232 10783-234 10783-236 10783-238 10783-240 10783-242 10783-244 10783-246 10783-248
OptiTDS™, Neural Tissue Dissociation System, CHI Scientific
Protein Extraction Protein Extraction Kits
OptiTDS™ is a proprietary Tissue Dissociation system which enzymatically digests extracellular connective fibers without harming the cells themselves
Stability/Storage: The reagents contained in this system are stable at room temperatures for normal shipping periods, but the kit should be used immediately or the package should be stored at -20oC upon arrival. The reagents contained in this kit are stable and effective for up to 6 months at -20oC.
Each OptiTDS™ system contains concentrated tissue dissociation enzymes and reagents which are added to an included buffer system to produce a working solution that is capable of treating 3-5 g of tissue. Each tissue dissociation system is customized to be tissue-specific and cell-specific to obtain the best results for each type of primary cell.