OptiTDS™, Mammary Tissue Dissociation System, CHI Scientific
Supplier: CHI Scientific
4-20321 4-20322 4-20323 4-20324 4-20325 4-20326 4-20327
10786-184EA 601.46 CAD
10786-184 10786-186 10786-188 10786-190 10786-192 10786-194 10786-196
OptiTDS™, Mammary Tissue Dissociation System, CHI Scientific
Protein Extraction Protein Extraction Kits
OptiTDS™ is a proprietary Tissue Dissociation system which enzymatically digests extracellular connective fibers without harming the cells themselves
Stability/Storage: The reagents contained in this system are stable at room temperatures for normal shipping periods, but the kit should be used immediately or the package should be stored at -20oC upon arrival. The reagents contained in this kit are stable and effective for up to 6 months at -20oC.
Each OptiTDS™ system contains concentrated tissue dissociation enzymes and reagents which are added to an included buffer system to produce a working solution that is capable of treating 3-5 g of tissue. Each tissue dissociation system is customized to be tissue-specific and cell-specific to obtain the best results for each type of primary cell.