OptiTDS™, Adipose/Fat Tissue Dissociation System, CHI Scientific
Supplier: CHI Scientific
4-20191 4-20192 4-20193 4-20194 4-20195
10786-070EA 601.46 CAD
10786-070 10786-072 10786-074 10786-076 10786-078
OptiTDS™, Adipose/Fat Tissue Dissociation System, CHI Scientific
Protein Extraction Protein Extraction Kits
OptiTDS™ is a proprietary Tissue Dissociation system which enzymatically digests extracellular connective fibers without harming the cells themselves
Stability/Storage: The reagents contained in this system are stable at room temperatures for normal shipping periods, but the kit should be used immediately or the package should be stored at -20oC upon arrival. The reagents contained in this kit are stable and effective for up to 6 months at -20oC.
Each OptiTDS™ system contains concentrated tissue dissociation enzymes and reagents which are added to an included buffer system to produce a working solution that is capable of treating 3-5 g of tissue. Each tissue dissociation system is customized to be tissue-specific and cell-specific to obtain the best results for each type of primary cell.