TexWrite® Notebook Binder, Texwipe®

Supplier: Texwipe
TexWrite®
TX5810
TWTX5810CS 663.3 CAD
TWTX5810
TexWrite® Notebook Binder, Texwipe®
Binders and Indexes
Designed for cleanroom equipment manuals, operating procedures and work instructions, the TexWrite® three-ring binder features a durable polyethylene cover with a plastic ring mechanism

  • All-plastic construction ensures cleanliness and corrosion resistance for long life
  • For use with hole punched cleanroom paper
  • 100% metal free: Corrosion-resistant
  • Chemical resistant cover
  • Colour: white

The rings open quickly with a single-sliding tab for easy changes to binder contents.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR