KOGO™ TPA Cleanroom Tape
Supplier: Texwipe
TPA1048BL TPA1048CL TPA1048WH TPA2036WHC
TWTPA0596WHCS 243.9 CAD
TWTPA0596WH TWTPA1048BL TWTPA1048CL TWTPA1048GR TWTPA1048OR TWTPA1048WH TWTPA2036WHC TWTPA3016CL TWTPA3016WH TW-TPA2036WHC
KOGO™ TPA Cleanroom Tape
Tapes
These tapes are constructed of low density polyethylene backing with an acrylic adhesive
  • Easy identification, protection and storage
  • Reduces need for cutting or splicing tape with available width options
  • Holds to irregular shapes and creates proper seal/protection
These tapes are available in five colors and four widths. Also they are ideal for color coding and product identification, and its high conformability makes it well suited for sealing and packaging materials in critical environments.
This construction is low in halogens, leachable chlorides and heavy metals, providing users with the ideal cleanroom tape solution.
Ordering information: Tapes are available in press-and-seal cleanroom bags and also gamma irradiated tapes are available.