Elma Clean 260 Dip and Splash Gentle Ultrasonic Cleaning Solution, Elma
Supplier: Elma
8000073 8000029
10724-602EA 605.03 CAD
10724-602 10724-604
Elma Clean 260 Dip and Splash Gentle Ultrasonic Cleaning Solution, Elma
Cleaning Agents Detergents and Surface Cleansers
Neutral, biodegradable Elma clean 260 dip and splash gently cleans sensitive surfaces. The cleaning solution is easily rinsed off with water leaving no residue. Use in an ultrasonic cleaner or labware washer
  • Ultrasonic cleaning concentrate for sensitive metals, plastics, and glass
  • Removes coolant, grease, oil, dust, fingerprints, and lime soaps
  • Ideal for cleaning aluminum prior to anodizing or coating
  • Contains no silicates and rinses residue-free
Elma clean 260 dip and splash is highly concentrated and should be diluted to 1-2% with water before use.More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable Sustainable Packaging,Safer Human and Environmental Health