BugBuster® (primary amine-free) Extraction Reagent, MilliporeSigma
Supplier: MilliporeSigma
BugBuster (primary amine-free) Protein Extraction Reagent is a special formulation of BugBuster designed for applications where primary amines would interfere if present in the protein extract, such as protein immobilization or cross-linking.
- Designed for protein immobilization
- Ideally suited for metal dependent, protein extraction
- Amine-free
The PIPPS buffer used in BugBuster (primary amine-free) has a similar buffer capacity and pH range as the original Tris-buffered BugBuster, but will not complex metal ions, also making it ideally suited for extraction of metal dependent proteins.